Skip to Content
Merck
All Photos(1)

Key Documents

1.03094

Sigma-Aldrich

5-Dimethylaminonaphthalene-1-sulfonyl chloride

for biochemistry

Synonym(s):

5-Dimethylaminonaphthalene-1-sulfonyl chloride, Dansyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12ClNO2S
CAS Number:
Molecular Weight:
269.75
MDL number:
UNSPSC Code:
12352106
EC Index Number:
210-092-6
NACRES:
NA.21

Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (HPLC)

form

solid

mp

69.0-73.0 °C

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Cl)c1c2c(c(ccc2)N(C)C)ccc1

InChI

1S/C12H12ClNO2S/c1-14(2)11-7-3-6-10-9(11)5-4-8-12(10)17(13,15)16/h3-8H,1-2H3

InChI key

XPDXVDYUQZHFPV-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance of solution (clearness): passes test
Appearance of solution (color): passes test
Melting point: 72 - 74 °C
Self-fluorescence (Chinin): ≤ 1.0 ppb

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service