Principaux documents
Z721328
FAST slide incubation chambers
single well incubation chamber (for 1 Pad FAST Slides) 10/pk
Synonyme(s) :
hybridization chambers, incubation chambers, slide chambers
About This Item
Code UNSPSC :
41122600
Nomenclature NACRES :
NB.22
Conditionnement
pack of 10
Fabricant/nom de marque
Maine Mfg 10486137
Chambre, L × l
79 mm × 25.4 mm
Volume
600-700 μL
Puits, L × l × P
53 mm × 22 mm × 4 mm
Description générale
Whatman array incubation chambers are designed for protein microarray applications on FAST Slides. The chambers provide a convenient way to carry out binding reactions on protein microarrays.
Used in conjunction with the FAST Frame or Chip Clip slide holder, the incubation chambers have a secure gasket design forming a tight, leak-proof seal with the FAST Slides. Simply remove the incubation chamber when the reaction is finished. These reusable chambers are recommended for use at room temperature and elevated incubation temperatures up to 76ºC. Disposable Chamber Covers are also available to prevent splashing and evaporation during incubations.