Principaux documents
GBL204970
Grace Bio-Labs FlexWell slide tray
holds up to 4 slides
Synonyme(s) :
Slide store, slide holder
About This Item
Code UNSPSC :
41123302
Nomenclature NACRES :
NB.22
Matériaux
acrylic
Caractéristiques
Holds 4 x slides 76x26 mm
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Grace Bio-Labs 204970
Taille externe
85.4 mm × 127.6 mm
Description générale
FlexWell 4 slide tray holds up to 4 slides, outside tray dimentsion L 85.4 mm x W 127.6 mm
Informations légales
FlexWell is a trademark of Grace Bio-Labs, Inc.