Principaux documents
Z238686
PTFE stirrer shaft
shaft diam. 6 mm, L 400 mm
About This Item
Code UNSPSC :
41103800
Nomenclature NACRES :
NB.72
Matériaux
PTFE
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Cowie 005.540.1
Longueur
400 mm
Diam. de l'axe
6 mm
Description générale
With 6.5mm diamond-shaped button and steel core, order blades below.
Stirring shaft with steel core for optimal rigidity. Use with PTFE stirrer guides and for applications which do not require a guide. Unbreakable, scratch resistant, and usable up to 280°C.