Principaux documents
Z112151
Sigma-Aldrich
32mm Round cell window
NaCl, Drilled
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Matériaux
NaCl
Conditionnement
pkg of (contains 1 window)
Technique(s)
IR spectroscopy: suitable
Diam. × épaisseur
32 mm × 3 mm
Description générale
Cut and polished for demountable liquid and gas cell holders