Principaux documents
Z151688
Sigma-Aldrich
Polished IR crystal window
NaCl
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Matériaux
NaCl
Technique(s)
IR spectroscopy: suitable
Diam. × épaisseur
13 mm × 2 mm
Description générale
Use with magnetic holder (Z112488) for films, neat, or Nujol samples.