Principaux documents
SYNF454250A
Synthware Squibb-style separatory funnel with standard taper joints, PTFE stopcock and PTFE stopper
250 mL, joint: ST/NS 24/40, PTFE stopcock and glass stopper
Synonyme(s) :
Synthware F454250A, glass separatory funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F454250A
Capacité
250 mL
250 mL
Description générale
With 2 mm PTFE stopcock. Supplied with a glass stopper or PTFE stopper.
Informations légales
Synthware is a trademark of Kemtech America Inc.