Principaux documents
SYNF780250
Synthware Squibb separatory funnel with glass stopcock
250 mL, stopper size #22
Synonyme(s) :
Synthware F780250, glass separatory funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F780250
Capacité
250 mL
250 mL
Description générale
Supplied complete with a flask length glass stopper of the listed size.
Informations légales
Synthware is a trademark of Kemtech America Inc.