Principaux documents
SYNF321200L
Synthware pear shaped flask
200 mL, joint: ST/NS 14/20
Synonyme(s) :
Synthware F321200L, pear shape glass flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F321200L
Capacité
200 mL
200 mL
Description générale
Heavy wall design.
Informations légales
Synthware is a trademark of Kemtech America Inc.