Principaux documents
SYNA761420
Synthware flushing adapter with T-bore PTFE stopcock
joint: ST/NS 14/20
Synonyme(s) :
Synthware A761420, glass flushing adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A761420
Description générale
With 2 mm T-bore PTFE stopcock.
Informations légales
Synthware is a trademark of Kemtech America Inc.