Principaux documents
HAM201150
Hamilton® Gastight® syringe, general use, 1700 series
model, 1750, AccuDil (AD) end, volume 500 μL
About This Item
Code UNSPSC :
41122001
Nomenclature NACRES :
NB.03
Matériaux
Borosilicate glass/PTFE
Gastight syringe
Caractéristiques
autoclavable: no
model 1750, AccuDil (AD) end
Fabricant/nom de marque
Hamilton
Paramètres
20-50 °C temp. range (68-122 °F)
700 psig pressure
Volume
500 μL
Informations légales
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.