Merck
Z709956
Sartorius pipette tips
volume range 10-1000 μL, standard, racked, non-sterile
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Maximales Volumen:
1 mL
Volumenbereich:
10-1000 μL
Sterilität:
non-sterile
Leistungsmerkmale:
DNase free
RNase free
barrier: no
endotoxin free
Verpackung:
pack of 96 ea (Racked)
Hersteller/Markenname:
Sartorius 791000
Materialien
colorless
Sterilität
non-sterile
Leistungsmerkmale
DNase free
RNase free
barrier: no
endotoxin free
Verpackung
pack of 96 ea (Racked)
Hersteller/Markenname
Sartorius 791000
Maximales Volumen
1 mL
Volumenbereich
10-1000 μL
Allgemeine Beschreibung
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment by being compatible with the Optiload feature in Sartorius Picus Nxt, Picus, Tacta, Mline and Proline Plus pipettes.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Sonstige Hinweise
Browse all pipette tips
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)