Merck
EP5418707005
Eppendorf® Rotor FA-45-18-11
incl. lid, aerosol tight
About This Item
UNSPSC-Code:
41103909
NACRES:
NB.82
Leistungsmerkmale
to hold 18 x 1.5/2.0 mL tubes
incl. lid, aerosol tight
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 5418707005
Methode(n)
centrifugation: suitable
Kompatibilität
for use with centrifuge models 5418/5418R 
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG