CLS70165101
Corning® VISTA™ Petri Dish
dish diam. 100 mm × 15 mm
Synonym(e):
glass cell culture dish
Empfohlene Produkte
Materialien
round
soda-lime glass dishes with lids
Sterilität
non-sterile
Leistungsmerkmale
lid
reusable
Verpackung
case of 24
Hersteller/Markenname
Corning 70165-101
Schalendurchm.
100 mm × 15 mm
Verwandte Kategorien
Allgemeine Beschreibung
The thermal expansion of Soda Lime Glass is 93.5 x 10-7 cm/cm/°C. The normal service temperature is 110°C and the extreme service temperature is 460°C. These dishes are not designed for rapid thermal shock. The Corning® brand Petri Dishes should be viewed as a low cost alternative to Pyrex® brand Petri Dishes (Catalog CLS3160) which are made from low expansion (32.5 x 10-7 cm/cm/°C) borosilicate glass.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)