CLS3514
Corning® FiltrEX™ plates
polystyrene non-sterile, 100 ea
Synonym(e):
96 multiwell plate
Empfohlene Produkte
product name
Fluid Guard for Corning® 96 well FiltrEX™ plates, polystyrene, non-sterile
Materialien
polystyrene
Sterilität
non-sterile
Hersteller/Markenname
Corning 3514
Verwandte Kategorien
Allgemeine Beschreibung
The fluid guard is manufactured from thermoformed polystyrene, non-sterile and easily snaps to the base of the filter plate. The unique design enables the guard to be applied manually or robotically to the plate. The guard serves to protect laboratory furniture and equipment from hazardous or radioactive materials.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
FiltrEX is a trademark of Corning, Inc.