Merck
AXYMTS06C
MiniTube
individual, bulk pack, size 0.65 mL, pkg of 5x960tips/cs, polypropylene, non-sterile
Synonym(e):
centrifuge tubes
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
polypropylene
Sterilität
non-sterile
Hersteller/Markenname
Axygen Inc MTS-06-C
Corning
Verpackung
pkg of 5x960tips/cs
Größe
0.65 mL
Leistungsmerkmale und Vorteile
  • Ensures positive sample identification due to one-way lid design, in conjunction with α-numeric grid
  • Compatible with Tecan®, Beckman Coulter, ANSI/SLAS 1-2004, microplates, and other robotic workstations
  • DNase/RNase free and non-pyrogenic in nature
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Coulter is a trademark of Beckman Coulter, Inc.
Tecan is a registered trademark of Tecan Group AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)