Merck
Z677965
Finntip® Flex pipette tips
Finntip Flex 300, non-sterile
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Materialien
colorless tip
orange
Sterilität
non-sterile
Leistungsmerkmale
barrier: no
Verpackung
pack of 960 ea (10 racks of 96 tips)
Hersteller/Markenname
Finn 94060513
Maximales Volumen
300 μL
Volumenbereich
5-300 μL
Allgemeine Beschreibung
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Sonstige Hinweise
Browse all pipette tips
Rechtliche Hinweise
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)