Merck
Z125202
Ace Claisen Adapter
Distilling
Synonym(e):
distilling adapter, glass adapter, glass adapters
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Etikett
B
Verbindung
joint ST/NS24/40
Hersteller/Markenname
Ace Glass 513654
Allgemeine Beschreibung
Threaded nylon bushing for 7 mm O.D. thermometer.
product is shown on the right in the photo
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)