Merck
Z106429
Supelco
Schwarze Kunststoffkappen
Solid top, PTFE-faced rubber liner, thread for 13-425, pkg of 200 ea
Synonym(e):
Protective Plastic Caps
About This Item
UNSPSC-Code:
41122001
NACRES:
NB.11
Materialien
PTFE-faced rubber liner
black phenolic cap
Leistungsmerkmale
closure type black phenolic solid top cap
Verpackung
pkg of 200 ea
Fitting
thread for 13-425
Allgemeine Beschreibung
It is autoclavable, easy sterilization, is used as a closure type for the containers used to contain substances in order to prevent contamination.
These autoclavable caps are designed to ensure the ultimate in product safety.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)