Merck
SYNF701430M
Synthware fritted filter tube
joint: ST/NS 14/20, disc O.D. 30 mm, porosity: medium
Synonym(e):
Synthware F701430M, straight filter tube
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F701430M
Scheibe AD
30 mm
Allgemeine Beschreibung
Available with a choice of coarse or medium porosity fritted discs.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)