Merck
SYNF490100
Synthware storage flask with high vacuum valve and side arm
100 mL, valve size 0-4 mm
Synonym(e):
Synthware F490100, round bottom flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
round bottom
single-neck (high vacuum valve)
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F490100
Kapazität
100 mL
100 mL
Ventilgröße
0-4 mm
Allgemeine Beschreibung
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper( “umbrella”) to protect Viton® O-ring on shaft..
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)