Merck
SYNF214250
Synthware four-neck round-bottom flask with vertical side necks
capacity 250 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
four-neck
round bottom
Verbindung
center joint: ST/NS24/40
side joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F214250
Kapazität
250 mL
Allgemeine Beschreibung
With identical vertical side joints.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)