Merck
SYNC195173
Synthware chromatography column with with spherical joint and PTFE stopcock
joint: ST/NS 35/20, I.D. × L 13.4 mm × 305 mm
Synonym(e):
Synthware C195173, glass chromatograpy column
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Verbindung
joint: ST/NS35/20
Qualität
for chromatography
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C195173
ID × L
13.4 mm × 305 mm
Verwandte Kategorien
Allgemeine Beschreibung
Spherical joint used with pinch clamp makes quick and tight connection under pressure for flash chromatography. All Synthware columns are heavy wall designed to increase safety.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)