SYNA972440
Synthware™ distilling adapter with compression cap
joint: ST/NS 24/40, side arm angle: 75 deg
Synonym(e):
Synthware A972440, glass distillation adapter
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A972440
Allgemeine Beschreibung
With compression cap (RC009) and Viton® O-ring for forming a vacuum tight seal of plain thermometer.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)