Merck
SYNA611420
Synthware straight vacuum adapter with glass stopcock
male joint: ST/NS 14/20
Synonym(e):
Synthware A611420, glass adapter with stopcock
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
male joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A611420
Allgemeine Beschreibung
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)