Merck
SYNA381922
Synthware 90 degree vacuum adapter with PTFE stopcock
male joint: ST/NS 19/22
Synonym(e):
Synthware A381922, adapter, vacuum adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
male joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A381922
Allgemeine Beschreibung
With 8 mm or 10 mm O.D. hose connection and 2 mm PTFE stopcock.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)