Merck
CLS9998241
Corning® phenolic cap, PTFE liner
24-410 thread
Synonym(e):
black phenolic, phenolic caps, screw caps, Corning® 9998
About This Item
UNSPSC-Code:
41122500
NACRES:
NB.43
Materialien
PTFE liner
black phenolic cap
Hersteller/Markenname
Corning 9998-241
Höhe
~18.5 mm
Fitting
24-410 thread
Allgemeine Beschreibung
These reusable Corning caps are fabricated from specially formulated resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface to provide a highly inert sealing face.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)