CLS70781CN
Pyrex® serological pipettes, disposable, sterile, plugged
volume 1.0 mL, accuracy: 0.01 mL
Synonym(e):
Corning® 7078
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.04
Empfohlene Produkte
Materialien
borosilicate glass
Beschreibung
plugged
Sterilität
sterile
Hersteller/Markenname
Corning 7078-1CN
Messbereich Volumen
1.0 mL, accuracy: 0.01 mL
Kapazität
1 mL
Allgemeine Beschreibung
These sterile disposable Pyrex serological pipettes are designed for the larger-volume user. They are calibrated "to deliver" (TD) with blow-out and are packed to permit easy access to several pipettes at a time without contaminating the entire case. Similar to No. 7077, except multi-packed in plastic bags.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)