Skip to Content
Merck
All Photos(1)

Documents

SML2722

Sigma-Aldrich

MU140

≥98% (HPLC)

Synonym(s):

1-Methyl-4-(9-(2-phenyl-phenyl)-7-oxa-2-aza-bicyclo[4.3.0]nona-1,3,5,8-tetraen-3-yl)-1H-pyrazole, 3-(Biphenyl-2-yl)-5-(1-methyl-1H-pyrazol-4-yl)furo[3,2-b]pyridine, 3-[1,1′-Biphenyl]-2-yl-5-(1-methyl-1H-pyrazol-4-yl)-furo[3,2-b]pyridine, MU1210 inactive control, MU1210 negative control, VN270, VN339 inactive control, VN339 negative control

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H17N3O
CAS Number:
Molecular Weight:
351.40
UNSPSC Code:
41121813
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CN(N=C1)C=C1C2=CC=C(OC=C3C4=C(C5=CC=CC=C5)C=CC=C4)C3=N2

Biochem/physiol Actions

MU140 is a structure analog of and the recommended inactive (negative) control for MU1210, an inhibitor against cdc-like kinase (cdc2-like kinase) CLK1, CLK2, CLK4, and homeodomain-interacting protein kinase 2 (HIPK2). MU140 exhibits no inhbitory potency toward CLK1/2/3/4, DYRK1a/1b/2 or HIPK1/2/3 (IC50 >3 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Václav Němec et al.
Angewandte Chemie (International ed. in English), 58(4), 1062-1066 (2018-12-21)
Reported is the identification of the furo[3,2-b]pyridine core as a novel scaffold for potent and highly selective inhibitors of cdc-like kinases (CLKs) and efficient modulators of the Hedgehog signaling pathway. Initially, a diverse target compound set was prepared by synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service