Skip to Content
Merck
All Photos(1)

Key Documents

820032

TOYOPEARL® AF-Heparin HC-650M Bulk Media

matrix active group sulfated glycosaminoglycan, bottle of 1 L, 65 μm particle size

Synonym(s):

TOYOPEARL® AF-Heparin HC-650M Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500

description

Adsorption capacity (> 5mg/mL (antithrombin III))
Phase filter (Heparin)
Shipping solvent (20% (v/v) ethanol)

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 20032

parameter

≤3 bar pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

sulfated glycosaminoglycan

particle size

65 μm

pore size

1000 Å

pH

5-10

capacity

>5 g/L adsorption capacity (antithrombin III)

separation technique

affinity

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl AF-Heparin HC-650 resin is a new high capacity affinity resin with an Antithrombin III (AT III) dynamic binding capacity of approximately 4 mg/mL. Due to its polyanionic nature heparin interacts with a wide range of biomolecules including plasma components, lipoprotein lipase, collagenase, and DNA polymerase

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service