
TOYOPEARL® AF-Heparin HC-650M Bulk Media

Synonym(s):
TOYOPEARL® AF-Heparin HC-650M Bulk Media
Products
Manufacturer
pH Value
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing