Skip to Content
Merck
All Photos(1)

Key Documents

AB15468

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody

CHEMICON®, chicken polyclonal

Synonym(s):

ChAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

product name

Anti-Choline Acetyltransferase Antibody, Chemicon®, from chicken

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Chat(12647)

Specificity

Choline acetyltransferase [ChAT]
Other species have not yet been tested. It is expected that the antibody will also react with human due to immunogen sequence similarity.

Immunogen

Synthetic peptide corresponding to human/mouse ChAT protein.

Application

Anti-Choline Acetyltransferase Antibody is an antibody against Choline Acetyltransferase for use in IC, IH.
Immunohistochemistry: 1:1,000-1:2,000 using 2% paraformaldehyde fixed tissue.

Immunocytochemistry: 1:1,000-1:2,000 using 2% paraformaldehyde fixed cells.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Physical form

ImmunoAffinity Purified
Liquid in PBS, pH 7.2 with 0.02% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SooHyun Kim et al.
Journal of neuroendocrinology, 32(1), e12830-e12830 (2020-01-17)
Rapid nonclassical effects of 17β-oestradiol (E2 ) on intracellular signalling have been identified in the basal forebrain, although the extent to which these actions may be different in males and females is unknown. Previous work has shown that E2 rapidly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service