Skip to Content
Merck
All Photos(1)

Key Documents

687839

Sigma-Aldrich

3-Nitro-4-hydroxyphenyl-13C6 acetic acid

99 atom % 13C, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C2H7NO5
Molecular Weight:
203.10
UNSPSC Code:
12352106
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

liquid

mp

146-148 °C

mass shift

M+6

SMILES string

O[13C]1=[13C][13C]=[13C](CC(O)=O)[13C]=[13C]1[N+]([O-])=O

InChI

1S/C8H4NO5/c10-7-2-1-5(4-8(11)12)3-6(7)9(13)14/h10H,4H2,(H,11,12)/i1+1,2+1,3+1,5+1,6+1,7+1

InChI key

COVPALHZVMEROO-JTZKEMBVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service