Y0002139
Zolpidem for system suitability
CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Zolpidem, N,N,6-Trimethyl-2-(4-methylphenyl)-imidazo[1,2-a]pyridine-3-acetamide
About This Item
Recommended Products
Quality Level
API family
zolpidem
manufacturer/tradename
EDQM
storage temp.
2-8°C
SMILES string
CC1=CN2C(C=C1)=NC(C3=CC=C(C)C=C3)=C2CC(N(C)C)=O
InChI
1S/C19H21N3O/c1-13-5-8-15(9-6-13)19-16(11-18(23)21(3)4)22-12-14(2)7-10-17(22)20-19/h5-10,12H,11H2,1-4H3
InChI key
ZAFYATHCZYHLPB-UHFFFAOYSA-N
Gene Information
human ... GABRA1(2554) , GABRB1(2560) , GABRG2(2566)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service