Skip to Content
Merck
All Photos(1)

Key Documents

CRM44892

Supelco

Glycerol solution

certified reference material, ASTM® D6584, 500 μg/mL in pyridine

Synonym(s):

Glycerol solution, Biodiesel impurity, 1,2,3-Propanetriol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH · aq
CAS Number:
Molecular Weight:
92.09 (anhydrous basis)
Beilstein:
635685
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

ASTM® D6584 Glycerin solution, certified reference material, 500 μg/mL in pyridine

grade

certified reference material
TraceCERT®

Agency

ASTM® D6584

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

500 μg/mL in pyridine

technique(s)

gas chromatography (GC): suitable

bp

182 °C/20 mmHg (lit.)

density

1.252 g/mL at 25 °C (lit.)

application(s)

petroleum

format

single component solution

storage temp.

2-8°C

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service