Skip to Content
Merck
All Photos(1)

Key Documents

48260-U

Supelco

4,4′-Dichlorobiphenyl solution

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 508

feature

standard type surrogate

packaging

ampule of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

InChI

1S/C12H8Cl2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H

InChI key

YTBRNEUEFCNVHC-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Chu et al.
Water research, 36(9), 2187-2194 (2002-07-11)
Surfactant and organic solvents have individually been shown useful in assisting the solubilization of hydrophobic organics out of contaminated soil and promoting UV-induced photodecay at a succeeding treatment process. A newly proposed process is to use the mixtures of surfactant
L van Bree et al.
Toxicology, 42(2-3), 259-274 (1986-12-15)
Both 4,4'-dichlorobiphenyl (4,4'-DCB) and 3-methylcholanthrene (3-MC) caused a substantial increase of phenacetin-induced hepatic glutathione (GSH) depletion, whereas phenobarbital (PB) had no effect, suggesting that 4,4'-DCB possesses cytochrome P-448 inducing activity. The O-deethylation of phenacetin by liver microsomes from control and
Uptake of 4-chlorobiphenyl and 4,4'-dichlorobiphenyl in six species of plant tissue cultures.
K Yamamoto et al.
Bulletin of environmental contamination and toxicology, 28(6), 728-732 (1982-06-01)
R Mavoungou et al.
The Science of the total environment, 101(3), 263-268 (1991-01-15)
Anaerobic cultures containing 4,4'-dichlorobiphenyl (4,4'CB) were inoculated with various environmental samples. The degradation of the substrate was followed by gas chromatographic analysis. Three of the cultures tested showed an ability to dehalogenate the substrate, as judged by the presence of
D Dietrich et al.
Applied and environmental microbiology, 61(11), 3904-3909 (1995-11-01)
The white rot fungus Phanerochaete chrysosporium has demonstrated abilities to degrade many xenobiotic chemicals. In this study, the degradation of three model polychlorinated biphenyl (PCB) congeners (4,4'-dichlorobiphenyl [DCB], 3,3',4,4'-tetrachlorobiphenyl, and 2,2',4,4',5,5'-hexachlorobiphenyl) by P. chrysosporium in liquid culture was examined. After

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service