Z637203
Mucasol™ universal detergent
volume 5 L
Synonym(s):
Mucasol cleaner, Multi-purpose detergent
About This Item
Recommended Products
manufacturer/tradename
BRAND 44005
volume
5 L
5000 mL
Looking for similar products? Visit Product Comparison Guide
General description
The universal cleaning agent for labware and instruments made of glass, porclain, plastics, rubber and metal instruments. Items are cleaned within 10 to 30 minutes, without mechanical help. Mucasol replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, non-aggressive and biodegradable.
Prolonged exposure to zinc and non-anodized aluminum surfaces should be avoided. Also suitable for cleaning by wiping.
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service