Key Documents
Z376957
Sigma-Aldrich
Gel Handler gel supports
sheet W × L × thickness 8 1/2 in. × 11 in. × 5 mil (0.125 mm)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.51
Pricing and availability is not currently available.
packaging
pkg of 10 sheets
sheet W × L × thickness
8 1/2 in. × 11 in. × 5 mil (0.125 mm)
General description
UV-transparent polyolefin Gel-Handler sheets easily slide under a gel resting in solution for transport to bench or transilluminator. The sheet is sturdy, but it conforms to the shape of the gel, reducing the trapping of air bubbles. Sheets are chemically inert, autoclavable and reusable.
May be cut to size with scissors.
Legal Information
Gel Handler is a trademark of C.B.S. Scientific Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)