Skip to Content
Merck
All Photos(1)

Key Documents

SRP6111

Sigma-Aldrich

SlyD from Escherichia coli

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

FKBP-type peptidyl prolyl cis-trans isomerase SlyD, Histidine-rich protein, Metallochaperone SlyD, Sensitivity to lysis protein D, WHP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Escherichia coli

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

21 kDa (196 aa, 1-196 aa)

packaging

pkg of 100 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Escherichia coli K12 ... slyD(947859)

General description

SlyD is a putative folding helper protein from the Escherichia coli cytosol, which consists of an N-terminal prolyl isomerase domain of the FKBP type and a presumably unstructured C-terminal tail. It is involved in the biosynthesis of the metal cluster in the [NiFe]-hydrogenase enzymes, and exhibits several activities including that of a peptidyl-prolyl isomerase.

Physical form

1 mg/mL solution in 20 mM Tris-HCl (pH 7.5).

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MKVAKDLVVS LAYQVRTEDG VLVDESPVSA PLDYLHGHGS LISGLETALE GHEVGDKFDV AVGANDAYGQ YDENLVQRVP KDVFMGVDEL QVGMRFLAET DQGPVPVEIT AVEDDHVVVD GNHMLAGQNL KFNVEVVAIR EATEEELAHG HVHGAHDHHH DHDHDGCCGG HGHDHGHEHG GEGCCGGKGN GGCGCH

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service