Skip to Content
Merck

Skip To

SML3312

SRI-41315

≥95% (HPLC), readthrough agent, powder

Synonym(s):

2-Cyclobutyl-10-methyl-3-phenylpyrimido[4,5-b]quinoline-4,5(3H,10H)-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H19N3O2
CAS Number:
Molecular Weight:
357.41
UNSPSC Code:
51111800
NACRES:
NA.32
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

SRI-41315, ≥95% (HPLC)

Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C1N(C2=CC=CC=C2)C(C3CCC3)=NC4=C1C(C5=CC=CC=C5N4C)=O

Biochem/physiol Actions

Cell penetrant and potent readthrough agents that suppress translation termination at premature termination codon
SRI-41315 is a cell penetrant and potent readthrough agents that suppress translation termination at premature termination codon and allows translation elongation to continue in the correct reading frame and generate a full-length protein. SRI-41315 induces the translational pause that occurs at premature termination codons by depleting eRF1 protein levels through a proteasome-mediated degradation pathway. SRI-41315 restores CFTR expression and function in in immortalized and primary human bronchial epithelial cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML3766HPA019004HPA073666
description

≥95% (HPLC)

description

≥98% (HPLC)

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

form

powder

form

powder

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

-

assay

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

-10 to -25°C

storage temp.

room temp

storage temp.

−20°C

storage temp.

−20°C

storage condition

desiccated

storage condition

-

storage condition

-

storage condition

-

solubility

DMSO: 2 mg/mL, clear (Warmed)

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

-

solubility

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters