Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502626

Sigma-Aldrich

Anti-DNA Polymerase ζ, N-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

DNA polymerase ζ catalytic subunit, DPOLZ, DPOZ, POLZ, REV3L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 352 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... REV3L(5980)

Related Categories

General description

Anti-DNA Polymerase ζ antibody detects endogenous levels of total DNA polymerase ζ protein.
The REV3L (reversionless 3-like) gene is mapped to human chromosome 6q21.[1] It codes for a ~350 kDa protein DNA polymerase ζ which localizes to mitochondria. [2]

Immunogen

The antiserum was produced against synthesized peptide derived from human DNA Polymerase zeta.

Immunogen Range: 231-280

Biochem/physiol Actions

REV3L (reversionless 3-like) is predominantly involved in DNA damage response, induced by ultraviolet radiation and chemical damaging agents such as cisplatin.[3][2] The encoded protein DNA Polymerase ζ catalyzes the translesion synthesis (TLS) with less stringent and lower processivity.[4] Loss of gene function is embryonically lethal. Upregulation of REV3L results in spontaneous mutations.[2]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

REV3L, the catalytic subunit of DNA polymerase ?, is involved in the progression and chemoresistance of esophageal squamous cell carcinoma.
Zhu X
Oncology Reports, 35(3), 1664-1670 (2016)
Loss of DNA polymerase zeta causes chromosomal instability in mammalian cells.
Wittschieben JP
Cancer Research, 66(1), 134-142 (2006)
Knockdown of REV3 synergizes with ATR inhibition to promote apoptosis induced by cisplatin in lung cancer cells.
Jiang HG
Journal of Cellular Physiology, 232(12), 3433-3443 (2017)
Tris-(2,3-Dibromopropyl) Isocyanurate, a New Emerging Pollutant, Impairs Cognition and Provokes Depression-Like Behaviors in Adult Rats.
Ye L
PLoS ONE, 10(10), 1-16 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service