Key Documents
M9064
Sigma-Aldrich
Thumbs-Up microtubes
volume 1.5 mL
Synonym(s):
plastic vials
About This Item
UNSPSC Code:
41121702
NACRES:
NB.22
Pricing and availability is not currently available.
volume
1.5 mL
General description
Saves fingernails from breaking and cracking. Opens with a flip of a thumb. Reduces sample contamination. Fast and easy removal from Rotors. Inner lid structure allows for pre-loading of reactants. RNase and DNase free. Fully autoclavable propylene. RCF rating of 17,000 × g. Can withstand temperatures ranging from - 196°C to 130°C.
Legal Information
Thumbs-Up is a trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)