Skip to Content
Merck
All Photos(3)

Key Documents

K1769

Sigma-Aldrich

Anti-KTN1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CG-1 antigen, Anti-Kinectin 1 (kinesin receptor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~150 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2-5 μg using lysate of Raji cells
indirect immunofluorescence: 2-5 μg/mL using human HeLa cells
western blot: 1-2 μg/mL using whole extract of human HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KTN1(3895)

General description

Kinectin (KTN1) protein is mainly located in the endoplasmic (ER) reticulum and is a large integral protein. This protein is made up of an N-terminal transmembrane domain and a C-terminal kinesin-binding domain. The KTN1 gene is mapped on the human chromosome at 14q22.3.

Specificity

Anti-KTN1 recognizes human KTN1.

Application

Anti-KTN1 antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Kinectin (KTN1) protein coheres to kinesin and is involved in vesicle transport on microtubules. This protein is also related to fibronectin-integrin adhesion sites.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derrek P Hibar et al.
Nature, 520(7546), 224-229 (2015-01-22)
The highly complex structure of the human brain is strongly shaped by genetic influences. Subcortical brain regions form circuits with cortical areas to coordinate movement, learning, memory and motivation, and altered circuits can lead to abnormal behaviour and disease. To
Huan Tran et al.
Journal of cell science, 115(Pt 10), 2031-2040 (2002-04-26)
Integrin receptors mediate the formation of adhesion complexes and play important roles in signal transduction from the extracellular matrix. Integrin-based adhesion complexes (IAC) contain proteins that link integrins to the cytoskeleton and recruit signaling molecules, including vinculin, paxillin, focal adhesion
Lee-Lee Ong et al.
The Journal of biological chemistry, 281(44), 33621-33634 (2006-09-05)
Kinectin is an integral membrane protein with many isoforms primarily found on the endoplasmic reticulum. It has been found to bind kinesin, Rho GTPase, and translation elongation factor-1delta. None of the existing models for the quaternary organization of the elongation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service