Skip to Content
Merck
All Photos(1)

Key Documents

EPB75-SQ

Eppendorf® Freezer Cardboard Storage Box

H 76 mm (3 in.) , tall box, white

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NB.12
Pricing and availability is not currently available.

material

cardboard

manufacturer/tradename

Eppendorf® B75-SQ (Global)

technique(s)

cryopreservation: suitable

H

76 mm (3 in.) , tall box

color

white

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service