Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C26H26N4O2S2 · 2HCl
CAS Number:
Molecular Weight:
563.56
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
storage temp.
−20°C
SMILES string
Cl.Cl.N[C@H](CSSC[C@H](N)C(=O)Nc1ccc2ccccc2c1)C(=O)Nc3ccc4ccccc4c3
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.