Skip to Content
Merck
All Photos(2)

Key Documents

10823

Supelco

trans-11-Eicosenoic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H38O2
CAS Number:
Molecular Weight:
310.51
Beilstein:
1727314
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCCCC(O)=O

InChI

1S/C20H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h9-10H,2-8,11-19H2,1H3,(H,21,22)/b10-9+

InChI key

BITHHVVYSMSWAG-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Mattarozzi et al.
Journal of chromatography. A, 1648, 462209-462209 (2021-05-18)
Two commercial stationary phases allowing both reversed phase mechanism and anion-exchange with different selectivity, i.e. CSH C18 and Atlantis PREMIER BEH C18 AX, were tested for the separation of a complex mixture of 21 fatty acids (FAs) encompassing saturated medium-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service