Skip to Content
Merck
All Photos(1)

Key Documents

R04586

Sigma-Aldrich

Formalin

ready-to-use solution, Harleco®, 10%

Synonym(s):

NBF Fixative, Formaldehyde solution 10%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352501
application(s):
diagnostic assay manufacturing
hematology
histology
Pricing and availability is not currently available.

form

ready-to-use solution

Quality Level

manufacturer/tradename

Harleco®

concentration

10%

color

colorless

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

General description

Formalin
R04586-76 1L Poly Bottle
HARLECO® Ready-to-use fixing solution, 10% Neutral Buffered Formalin (NBF)

Application

Click here for printer friendly instructions for adapting a hand pump to the 20L HDPE drum.
HARLECO® Ready-to-use fixing solution, 10% Neutral Buffered Formalin (NBF)

Components

EMD No. R04586-45 contains 450mL in a 1L Poly Jar

EMD No. R04586-01 is 2 x 10L

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service