Skip to Content
Merck
All Photos(1)

Key Documents

8.41003

Sigma-Aldrich

1,4-Diethylbenzene

for synthesis

Synonym(s):

1,4-Diethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14
CAS Number:
Molecular Weight:
134.22
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-265-2
Pricing and availability is not currently available.

vapor pressure

1.45 hPa ( 25 °C)

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

430 °C

expl. lim.

0.8 % (v/v)

bp

182-184 °C/1013 hPa

mp

-43 °C

transition temp

flash point 55 °C

solubility

24.8 mg/L

density

0.862 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H14/c1-3-9-5-7-10(4-2)8-6-9/h5-8H,3-4H2,1-2H3

InChI key

DSNHSQKRULAAEI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.860 - 0.865
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F

Flash Point(C)

56 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service