Skip to Content
Merck
All Photos(1)

Key Documents

GF99483126

Brass alloy, Cu63Zn37

1000mm tube, 1.5mm outside diameter, 0.25mm wall thickness, 1mm inside diameter, as drawn

Synonym(s):

CU027200

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
30102010
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 994-831-26

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service