Skip to Content
Merck
All Photos(1)

Key Documents

CDS000335

Sigma-Aldrich

3-Hydroxypropanoic acid solution

30% in water, AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

concentration

30% in water
30% in water

SMILES string

OC(CCO)=O

InChI

1S/C3H6O3/c4-2-1-3(5)6/h4H,1-2H2,(H,5,6)

InChI key

ALRHLSYJTWAHJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hidetoshi Morita et al.
DNA research : an international journal for rapid publication of reports on genes and genomes, 15(3), 151-161 (2008-05-20)
Lactobacillus reuteri is a heterofermentative lactic acid bacterium that naturally inhabits the gut of humans and other animals. The probiotic effects of L. reuteri have been proposed to be largely associated with the production of the broad-spectrum antimicrobial compound reuterin
Katharina Anna Lindlbauer et al.
New biotechnology, 34, 32-39 (2016-10-23)
This study investigates potential limitations of 1,3-propanediol formation by Lactobacillus diolivorans. Particular focus is given to enhanced glycerol utilization as well as the elimination of by-product formation. The key aspect is a modulation of the redox household by process engineering
Miranda G M Kok et al.
Talanta, 197, 49-58 (2019-02-18)
Volumetric absorptive microsampling (VAMS) enables the collection of small and accurate quantities of biological fluids. Therefore, this sampling technique is of great interest for volume-limited samples or serial collection of samples. In this study, we examined the potential of VAMS
Shinobu Sato et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 35(1), 85-90 (2018-11-06)
A peptide-oligonucleotide conjugate (1) was synthesized by the attachment of FAM, TAMRA, and biotin moieties to a telomere DNA sequence of 5'-TAG GGT TAG GGT TAG GGT TAG GG-3'. This conjugate was induced to be an anti-parallel structure in the
Andrew J Loder et al.
Metabolic engineering, 38, 446-463 (2016-10-25)
The 3-hydroxypropionate/4-hydroxybutyrate (3HP/4HB) cycle fixes CO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service